Order Entry
ContactUsLinkComponent
Human Recombinant PAI2 (from E. coli)
Human Recombinant PAI2 (from E. coli)
Catalog #: 10072-644
Supplier:  Prosci
CAS Number:  
Human Recombinant PAI2 (from E. coli)
Catalog #: 10072-644
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.002 mg
  • Biological activity:
    Determined by its inhibitory effect against single chain tPA induced cleavage of a chromogenic substrate in Imidazole Buffer at 37˚C. Half maximal inhibition against 1.0 μg/mL of single chain tPA was obtained at a concentration of 1.0 μg/mL
  • Protein synonyms:
    PLANH2|Plasminogen activator inhibitor 2|PAI2|PAI|HsT1201|Monocyte Arg-serpin|PAI-2
  • Protein/peptide name:
    PAI2
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-644
  • Supplier No.:
    40-127

Specifications

About this item

PAI-2 is an inhibitory serpin expressed mainly in keratinocytes, activated monocytes, and placental trophoblasts. It exists predominantly as a 47 kDa nonglycosylated intracellular protein which can be induced to be secreted as 60 kDa glycoprotein. The physiological relevance of this property, which is neither a consequence of any mutation in the PAI-2 gene nor associated with any known disorder, is still unclear. However, it appears that the formation of intracellular dormant polymers may be important for the controlled release of the inhibitor from PAI-2 producing cells. Plasma levels of PAI-2 are usually low or undetectable, except during pregnancy and in some forms of monocytic leukemia. Secretion of PAI-2 from the placenta normally occurs during the third trimester of pregnancy and accounts for the dramatic increase in PAI-2 levels (up to 250 ng/mL), which are maintained at these levels until postpartum, and then rapidly decline. In addition to its vital role in protecting the placenta from degradation by uPA and/or uPA-activated proteases, PAI-2 has been shown to be essential for the prevention of metastatic spread of neck, lung and breast cancers. The beneficial effect of PAI-2 seen in these studies is presumed to stem from its ability to inhibit uPA-dependent cell dissemination. PAI-2 has also been reported to inhibit keratinocyte proliferation, and to participate in the innate immune response during viral infection. Recombinant human PAI-2 is a 415-residue nonglycosylated protein.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought