Order Entry
ContactUsLinkComponent
Human Recombinant TWEAK Receptor (from E. coli)
Human Recombinant TWEAK Receptor (from E. coli)
Catalog #: 10072-918
Supplier:  Prosci
CAS Number:  
Human Recombinant TWEAK Receptor (from E. coli)
Catalog #: 10072-918
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to inhibit TWEAK - induced weak cell death of HT29 cells. The expected ED50 for this effect is 1.0 - 3.0 μg/mL.
  • Protein synonyms:
    FGF-inducible 14|CD266|FN14|Fibroblast growth factor-inducible immediate-early response protein 14|TWEAKR|Tumor necrosis factor receptor superfamily member 12A
  • Protein/peptide name:
    TWEAK Receptor
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-918
  • Supplier No.:
    40-312

Specifications

About this item

TWEAKR belongs to the TNF family of transmembrane proteins and contains a cytoplasmic "death domain", which can activated the cell's apoptotic machinery. It is expressed in the spleen, thymus, peripheral blood lymphocytes, colon, and small intestine. Signal transduction by TWEAKR can be activated by either the membrane anchored or the soluble TWEAK. Recombinant soluble TWEAKR is a 53 amino acid polypeptide (5.6 kDa) comprising the entire extracellular domain of the full length TWEAKR protein.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought