Order Entry
ContactUsLinkComponent
Human Recombinant TRAILR1 (soluble) (from E. coli)
Human Recombinant TRAILR1 (soluble) (from E. coli)
Catalog #: 10072-912
Supplier:  Prosci
CAS Number:  
Human Recombinant TRAILR1 (soluble) (from E. coli)
Catalog #: 10072-912
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    Biological activity was determined by its inhibitory effect of IL - 8 production in human PBMC by human sTRAIL/Apo2L. 95% inhibition (100 ng/mL of s TRAIL/Apo2L) was reached using a concentration of 100 ng/mL of sTRAIL Receptor - 1.
  • Protein synonyms:
    Death receptor 4|TRAILR1|DR4|Tumor necrosis factor receptor superfamily member 10A|TRAIL receptor 1|APO2|CD261|TRAILR-1
  • Protein/peptide name:
    TRAILR1 (soluble)
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-912
  • Supplier No.:
    40-309

Specifications

About this item

TRAIL Receptor-1/DR4 and TRAIL Receptor-2/DR5 belong to the TNFR superfamily of transmembrane proteins and contain a cytoplasmic "death domain, " which can activate the cell's apoptotic machinery. These receptors are activated by binding to either membrane anchored or soluble TRAIL/Apo2L. Recombinant human soluble TRAIL Receptor-1/DR4 is a 22.7 kDa protein (215 amino acid residues) consisting of the TNFR homologous, cysteine rich portion of the extracellular domain.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought