Order Entry
ContactUsLinkComponent
Mouse Recombinant ATG5 (from E. coli)
Mouse Recombinant ATG5 (from E. coli)
Catalog #: 10073-152
Supplier:  Prosci
CAS Number:  
ATG5 Recombinant Protein
Mouse Recombinant ATG5 (from E. coli)
Catalog #: 10073-152
Supplier:  Prosci
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Protein synonyms:
    hAPG5|APG5|APG5L|APG5-LIKE|ATG5 ASP
  • Protein/peptide name:
    ATG5
  • Purity:
    >95%
  • Molecular weight:
    34 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested applications:
    ELISA, Western Blot
  • Cat. no.:
    10073-152
  • Supplier No.:
    95-121

Specifications

About this item

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components (1,2). This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1 (3). ATG5, another member of the autophagy protein family, forms a conjugate with ATG12; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy (4). This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells (5).

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

1X PBS containing 10% glycerol

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought