Order Entry
ContactUsLinkComponent
Human Recombinant Visfatin (from E. coli)
Human Recombinant Visfatin (from E. coli)
Catalog #: 10072-618
Supplier:  Prosci
CAS Number:  
Human Recombinant Visfatin (from E. coli)
Catalog #: 10072-618
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    The ED50 was determined by the dose - dependant proliferation of the RPMI 8226 cells. The expected ED50 for this effect is 15.0 - 20.0 ng/mL.
  • Protein synonyms:
    VISFATIN|NAmPRTase|Nicotinamide phosphoribosyltransferase|PBEF1|PBEF|1110035O14Rik|Pre-B-cell colony-enhancing factor 1|VF
  • Protein/peptide name:
    Visfatin
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-618
  • Supplier No.:
    40-111

Specifications

About this item

Visfatin is a 55 kDa protein produced and secreted primarily by white adipose tissue. Recently, Visfatin was isolated from visceral fat deposits and shown to possess insulin-mimetic activity. Like insulin, Visfatin exerts hypoglycemic effects by interacting with the insulin receptor. The binding affinity of Visfatin for the insulin receptor is similar to that of insulin, but it does not compete with insulin, suggesting that the two proteins interact with different receptor sites. The circulating levels of Visfatin are much lower than those of insulin and are not affected by feeding, implying that the hypoglycemic effect of Visfatin may not be of physiological importance. The plasma Visfatin levels, like those of Leptin, correlate positively with the percent of body fat and increase during the development of obesity. Another similarity between Visfatin and Leptin is that their amino acid sequence is highly conserved across different mammalian species and shows no homology to any known protein. Receptors for both Leptin (Ob-R) and Visfatin (i.e. the insulin receptor) are expressed by neurons within the arcuate nucleus of the hypothalamus, a brain area that plays a pivotal role in the regulation of energy metabolism. Although the metabolic function of Visfatin is still unknown, it appears that this newly identified adipocytokine might play an important role, similar to that of Leptin, in the regulation of body weight, i.e. as an afferent signal reflecting excess body fat. Recombinant human Visfatin is a 52.5 kDa protein containing 465 amino acid residues (isoform 1).

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought