Order Entry
ContactUsLinkComponent
Human Recombinant CXCL8 (from E. coli)
Human Recombinant CXCL8 (from E. coli)
Catalog #: 10072-608
Supplier:  Prosci
CAS Number:  
Human Recombinant CXCL8 (from E. coli)
Catalog #: 10072-608
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to chemoattract human peripheral blood neutrophils using a concentration range of 25.0 - 150.0 ng/mL.
  • Protein synonyms:
    IL-8|MDNCF|Interleukin-8|LYNAP|NAP1|NAP-1|GCP1|MONAP|LECT|C-X-C motif chemokine 8|NAF|GCP-1|CXCL8|LUCT
  • Protein/peptide name:
    CXCL8
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-608
  • Supplier No.:
    40-105

Specifications

About this item

CXCL8 (IL-8) is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. CXCL8 (IL-8) chemoattracts and activates neutrophils. Recombinant human CXCL8 (IL-8) (endothelial-derived) is an 8.9 kDa protein containing 77 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought