Specifications
- Description:EMax® Plus Microplate Reader
- Cat. no.:10119-370
- Supplier No.:EMAX PLUS
Specifications
About this item
Robust, high value microplate reader designed to deliver research-grade results to any laboratory.
- Research-grade results
- Pre-defined protocols
- Eight standard filter modes
Running ELISAs and protein quantification is simpler than ever with the EMax® Plus Microplate Reader
EMax® Plus Microplate Reader benefits include, 8 filters for a variety of assays (covering visible range from 405–750nm), a compact instrument footprint, pre-defined protocols with SoftMax® Pro Software, and walk-up usability.
Standard features on the EMax® Plus Microplate Reader include the ability to measure both flat and round 96-well microplates, automatic lamp calibration prior to each reading (ensuring automatic measurements), powerful curve fitting protocols and statistical analysis tools, and integrated instrument control and data analysis with SoftMax® Pro Software. The software enables data visualization in 3D plots, gray scale, kinetic curve fits, and IC50 determinations.
In seconds, users can set up the reader with pre-defined protocols and run samples. With 8 standard filter modes that cover the entire visible range, walk-up usability, and SoftMax® Pro Software for data analysis, getting started is now simpler than ever.