To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:uridine phosphorylase 1
- Clonality:Polyclonal
- Gene ID:Q16831
- Host:Chicken
- Reactivity:Human
- Antigen symbol:UPP1
- Conjugation:Unconjugated
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:UPP1
- Storage buffer:Phosphate-Buffered Saline. No preservatives added.
- Molecular weight:19.2 kDa (calculated)
- Storage temperature:UPP1 antibody can be stored at 4 °C for short term (weeks). Long term storage should be at –20 °C. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:164-310
- Tested applications:Immunoaffinity Purified
- Purification:Immunoaffinity Purified
- Cat. no.:10071-710
- Supplier No.:XW-7924
Specifications
About this item
UPP1 catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate. The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis. It is involved in the pathway of nucleotide and deoxyribonucleotide catabolism.
UPP1 antibody can be used for the detection of UPP1 protein by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: UPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human