To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:hydroxyacylglutathione hydrolase
- Clonality:Polyclonal
- Gene ID:Q16775
- Host:Chicken
- Reactivity:Human,Rat,Mouse
- Antigen symbol:HAGH
- Conjugation:Unconjugated
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HAGH
- Storage buffer:Phosphate-Buffered Saline. No preservatives added.
- Molecular weight:28.9 kDa (calculated)
- Storage temperature:HAGH antibody can be stored at 4 °C for short term (weeks). Long term storage should be at –20 °C. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:110-260
- Tested applications:Immunoaffinity Purified
- Purification:Immunoaffinity Purified
- Cat. no.:10071-282
- Supplier No.:XW-7892
Specifications
About this item
FUNCTION: Thiolesterase that catalyzes the hydrolysis of S-D-lactoyl-glutathione to form glutathione and D-lactic acid.
CATALYTIC ACTIVITY: S-(2-hydroxyacyl)glutathione + H2O = glutathione + a 2-hydroxy carboxylate.
SUBUNIT: Monomer.
SIMILARITY: Belongs to the glyoxalase II family.
COFACTOR: Binds 2 zinc ions per subunit.
PATHWAY: Glyoxal pathway.
Hydroxyacylglutathione hydrolase antibody can be used for the detection of Hydroxyacylglutathione hydrolase by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: HAGH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat