To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, metabotropic 5
- Clonality:Polyclonal
- Gene ID:P31424
- Host:Rabbit
- Reactivity:Rat
- Antigen symbol:Grm5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Grm5
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:125/250
- Storage temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping temperature:4 °C
- Immunogen:mGluR 5/1a polyclonal antibody was raised against a peptide from the C-terminal region of mGluR5 and mGluR1a.
- Purification:Affinity Purified
- Cat. no.:10075-602
- Supplier No.:XPS-2018
Specifications
About this item
The metabotropic glutamate receptors (mGluRs) are key receptors in the modulation of excitatory synaptic transmission in the central nervous system. They are implicated in many forms of neural plasticity as well as learning and memory and drug abuse. Group I metabotropic glutamate receptors (consisting of mGluR1 and mGluR5) are G-protein-coupled neurotransmitter receptors that are localized in the perisynaptic region of the postsynaptic membrane. When activated, Group I mGluRs lead to stimulation of phospholipase and activation of Protein Kinase C. In contrast activation of Group II metabotropic receptors (mGluR2 and mGluR3) leads to inhibition of adenylate cyclase.
Metabotropic Glutamate Receptor 5/1a antibody allows for strong and specific immunolabeling of ~125k mGluR5 and mGluR1a and the ~250k mGluR5 and mGluR1a dimers in Western blots of rat brain at antibody dilution of 1:1000. Immunolabeling blocked by preadsorption of antibody with the C-terminal peptide immunogen. Applications include Dot Blots (DB), Immunohistochemistry (IHC) and Western Blots (WB). Rabbit anti-mGluR 5/1a recognizes both mGluR5 and mGluR1a in rat brain. When internally tested under ideal conditions the working dilutions were 1:500 for IHC and 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Grm5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat