To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, beta 3
- Clonality:Polyclonal
- Gene ID:P63079
- Host:Rabbit
- Reactivity:Human,Bovine,Rat
- Antigen symbol:Gabrb3
- Conjugation:Unconjugated
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Gabrb3
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:53
- Storage temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping temperature:4 °C
- Immunogen:GABAA Receptor b3 (CT) polyclonal antibody was raised against a fusion protein from the amino acid sequence representing the cytosolic loop of rat GABAA-Receptor b3-Subunit.
- Purification:Affinity Purified
- Cat. no.:10075-716
- Supplier No.:XPS-2012
Specifications
About this item
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA-Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date six alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha and beta subunits results in the expression of functional GABAA-Rs sensitive to GABA. However, coexpression of a gamma subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha subunits of the receptor. Lastly, phosphorylation of beta subunits of the receptor has been shown to modulate GABAA-R function.
GABAA Receptor antibody is specific for the 51 to 56k GABAA-Receptor ?3-Subunit in rat brain membrane fractions. Antibody dilutions and tissue load should be based on tissue type and expected expression level. Initial recommended range of dilutions: 1:1000 to 1:2000. Applications include Dot Blots (DB) and Western Blot (WB). Suitability for Immunohistochemistry (IHC) not yet determined. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Gabrb3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Bovine