To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, ionotropic, AMPA 1
- Clonality:Polyclonal
- Gene ID:P19490
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Gria1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.15 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Gria1
- Modification:Ser 845
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:100
- Storage temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping temperature:4 °C
- Immunogen:GluR1 (Ser845) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phospho-Ser845 of rat GluR1.
- Purification:Affinity Purified
- Cat. no.:10075-662
- Supplier No.:XPS-1013
Specifications
About this item
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5-methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of 4 distinct subunits (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR1 subunit is widely expressed throughout the nervous system. GluR1 is also potentiated by phosphorylation at Ser845. In addition, phosphorylation of this site has been linked to synaptic plasticity as well and learning and memory.
Immunolabeling of the GluR1 protein in western blots of rat brain is blocked by the Ser845 phosphopeptide used as antigen but not by the corresponding non-phosphopeptide. The antibody is purified by sequential chromatography on phospho- and non-phosphopeptide affinity columns. Antibody dilutions and tissue load should be based on tissue type and expected phosphorylation state. Initial recommended range of dilutions: 1:500 to 1:2000. Applications include Immunofluorescence (IF), Immunohistochemistry (IHC) and Western Blots (WB). Immunohistochemistry (IHC) has not yet been determined. Human, mouse and rat have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for IF, IHC and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Gria1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat