To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent protein kinase II alpha
- Clonality:Polyclonal
- Gene ID:P11275
- Host:Rabbit
- Reactivity:Xenopus,Human,Rat,Mouse
- Antigen symbol:Camk2a
- Conjugation:Unconjugated
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Camk2a
- Modification:Thr 286
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:50/60
- Storage temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping temperature:4 °C
- Immunogen:CaM Kinase II (Thr286) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acid residues surrounding the phosphorylated-Thr286 found in rat brain CaM Kinase II.
- Purification:Affinity Purified
- Cat. no.:10075-426
- Supplier No.:XPS-1000
Specifications
About this item
Calcium/calmodulin kinase II (CaM Kinase II) is multi-functional calcium and calmodulin-dependent protein kinase that mediates cellular responses to second messengers. Phosphorylation of Thr286 produces an autonomously active form of CaM Kinase II. The antibody was raised against a synthetic phosphopeptide corresponding to amino acid residues surrounding the phosphorylated-Thr286 found in rat brain CaM Kinase II.
Antibody dilutions and tissue load should be based on tissue type and expected phosphorylation state. Western blots of rat brain lysates with phospho- and dephosphopeptide competition were used to establish the phospho-specificity of the antibody. Applications include Dot Blot (DB) and Western Blot (WB). Dot Blots and Western Blots performed with each lot and working dilutions internally tested under ideal conditions for DB and WB were 1:1000. Suitability for Immunohistochemistry (IHC) has not been determined. Human, mouse, rat and Xenopus have 100% amino acid sequence identity with the antigen used to raise the antibody.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Camk2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus