To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor
- Clonality:Polyclonal
- Gene ID:P01375
- Host:Rabbit
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:TNF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TNF
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hTNF-alpha (human Tumor Necrosis Factor-alpha).
- Purification:Anti-hTNF-alpha specific antibody was purified by affinity chromatography employing immobilized hTNF-alpha matrix.
- Cat. no.:10097-646
- Supplier No.:XP-5282
Specifications
About this item
Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide stimulated macrophages, and causes tumor necrosis when injected into tumour bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hTNF-? (0.25 ng/mL), a concentration of 0.012 - 0.014 µg/mL of this antibody is required. ELISA: To detect hTNF-? by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hTNF-?. Sandwich: To detect hTNF-? by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human TNF-? (XP-5282Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTNF-?. Western Blot: To detect hTNF-? by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTNF-? is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-hTNF-alpha specific antibody was purified by affinity chromatography employing immobilized hTNF-alpha matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human