To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 17
- Clonality:Polyclonal
- Gene ID:Q92583
- Host:Rabbit
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CCL17
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL17
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hTARC (human Thymus and Activation Regulated Chemokine).
- Purification:Anti-hTARC specific antibody was purified by affinity chromatography employing immobilized hTARC matrix.
- Cat. no.:10097-634
- Supplier No.:XP-5279
Specifications
About this item
Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. TARC displays chemotactic activity for T lymphocytes, but not monocytes or granulocytes. TARC binds to chemokine receptors CCR4 and CCR8. This chemokine plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hTARC (100.00 ng/mL), a concentration of 20.0 - 30 µg/mL of this antibody is required. ELISA: To detect hTARC by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hTARC. Sandwich: To detect hTARC by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human TARC (XP-5279Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hTARC. Western Blot: To detect hTARC by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hTARC is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-hTARC specific antibody was purified by affinity chromatography employing immobilized hTARC matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CCL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human