To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 13
- Clonality:Polyclonal
- Gene ID:Q99616
- Host:Goat
- Reactivity:Human
- Antigen symbol:CCL13
- Conjugation:Biotin
- ELISA:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL13
- Shipping temperature:RT
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hMCP-4 (human MCP-4).
- Purification:Anti-hMCP-4 specific antibody was purified by affinity chromatography and then biotinylated.
- Cat. no.:10097-528
- Supplier No.:XP-5225BT
Specifications
About this item
MCP4 is a member of the b-chemokine family (C-C) of Cytokines. Like Eotaxin, MCP4 is a powerful eosinophil chemo-attractant. Additionally, like MCP-3, MCP-4 elicits chemo-attraction of monocytes and T lymphocytes
ELISA: To detect hMCP-4 by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hMCP-4. Sandwich: To detect hMCP-4 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human MCP-4 (XP-5225) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hMCP-4. Western Blot: To detect hMCP-4 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMCP-4 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-hMCP-4 specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CCL13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human