To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:interleukin 6
- Clonality:Polyclonal
- Gene ID:P08505
- Host:Rabbit
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:Il6
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Il6
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-6 (murine Interleukin-6).
- Purification:Anti-mIL-6 specific antibody was purified by affinity chromatography employing immobilized mIL-6 matrix.
- Cat. no.:10098-128
- Supplier No.:XP-5194
Specifications
About this item
IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. In addition, IL-6 appears to interact with IL2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 is a multifunctional 24 kDa protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL6 has also been implicated in regulating adipose mass.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of mIL-6 (0.1 ng/mL), a concentration of 0.012 -0.014 µg/mL of this antibody is required. ELISA: To detect mIL-6 by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant mIL-6. Sandwich: To detect mIL-6 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Murine IL-6 (XP-5194Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mIL-6. Western Blot: To detect mIL-6 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mIL-6 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-mIL-6 specific antibody was purified by affinity chromatography employing immobilized mIL-6 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Il6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse