To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:interleukin 10
- Clonality:Polyclonal
- Gene ID:P18893
- Host:Rabbit
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:Il10
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Il10
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-10 (murine IL-10).
- Purification:Anti-mIL-10 specific antibody was purified by affinity chromatography employing immobilized mIL-10 matrix.
- Cat. no.:10097-842
- Supplier No.:XP-5162
Specifications
About this item
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL. Human IL10 is a non glycosylated polypeptide consisting of 178 amino acids. There is 73% homology between the human and mouse IL10 proteins, however, the human IL10 acts on both human and mouse target cells, while the mouse IL10 has species specific activity. The cellular sources of IL10 are CD4+ T cells and T cell clones, thymocytes, B cells and B cell lymphomas, macrophages, mast cell lines and keratinocytes. IL10 will stimulate the growth of stem cells, mast cells and thymocytes. IL10 enhances cytotoxic T cell development, and costimulates B cell differentiation and immunoglobulin secretion. IL10 inhibits cytokine production by macrophages and suppresses macrophage class II MHC expression. The human IL10 gene is on human chromosome 1.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of mIL-10 (2.5 ng/mL), a concentration of 0.012- 0.014 µg/mL of this antibody is required. ELISA: To detect mIL-10 by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant mIL-10. Sandwich To detect mIL-10 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Murine IL-10 (XP-5162Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mIL-10. Western Blot: To detect mIL-10 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mIL-10 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-mIL-10 specific antibody was purified by affinity chromatography employing immobilized mIL-10 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Il10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse