To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:interferon gamma
- Clonality:Polyclonal
- Gene ID:P01581
- Host:Goat
- Neutralisation:Yes
- Reactivity:Rat
- Antigen symbol:Ifng
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Ifng
- Shipping temperature:RT
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant rIFN-gamma (rat Interferon-gamma).
- Purification:Anti-rIFN-gamma specific antibody was purified by affinity chromatography employing immobilized rIFN-gamma matrix.
- Cat. no.:10097-826
- Supplier No.:XP-5157
Specifications
About this item
IFN gamma receptor beta is part of the receptor for interferon gamma. This class II cytokine receptor pairs with CDw119 to form the IFN gamma receptor and is an integral part of the IFN gamma signal transduction pathway. CDw119 serves as the IFN gamma binding chain and associates with the IFN gamma beta chain which is required for receptor signaling. The extracellular portion of both the IFN gamma receptor alpha and beta chains must be species matched. The IFN gamma receptor beta chain is expressed on T and B cells, NK cells, monocytes/ macrophages, and fibroblasts. Binding of IFN gamma induces receptor dimerization, internalization, Jak1 and Jak2 protein kinase activation and, ultimately, STAT1 activation. It is also likely to interact with GAF. IFN gamma initiates and regulates a variety of immune responses and is required for signal transduction. Contains 2 fibronectin type III domains. Defects in IFN gamma Receptor beta are a cause of mendelian susceptibility to mycobacterial disease (MSMD), a rare condition that confers predisposition to illness caused by several mycobacteria strains
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of rIFN-gamma (0.3 ng/mL), a concentration of 0.006 - 0.01 µg/mL of this antibody is required. ELISA: To detect rIFN-gamma by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant rIFN-gamma. Sandwich: To detect Rat IFN-? by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Rat IFN-? (XP-5157Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant Rat IFN-?. Western Blot: To detect rIFN-gamma by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant rIFN-gamma is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-rIFN-gamma specific antibody was purified by affinity chromatography employing immobilized rIFN-gamma matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Ifng
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat