To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:fibroblast growth factor 16
- Clonality:Polyclonal
- Gene ID:O43320
- Host:Goat
- Reactivity:Human
- Antigen symbol:FGF16
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:FGF16
- Shipping temperature:RT
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hFGF-16 (human Fibroblast Growth Factor-16).
- Purification:Anti-hFGF-16 specific antibody was purified by affinity chromatography employing immobilized hFGF-16 matrix.
- Cat. no.:10098-462
- Supplier No.:XP-5132
Specifications
About this item
ELISA:
To detect hFGF-16 by direct ELISA (using 100 ?L/well antibody solution) a concentration of at least 0.5 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hFGF-16.
Sandwich: To detect hFGF-16 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human FGF-16 (XP-5132Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hFGF-16.
Western Blot:
To detect hFGF-16 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hFGF-16 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FGF16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human