To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 21
- Clonality:Polyclonal
- Gene ID:O00585
- Host:Rabbit
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CCL21
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL21
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hExodus-2 (human Exodus-2).
- Purification:Anti-hExodus-2 specific antibody was purified by affinity chromatography employing immobilized hExodus-2 matrix.
- Cat. no.:10098-446
- Supplier No.:XP-5128
Specifications
About this item
Exodus 2 is one of several CC cytokine genes clustered on the p arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterised by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hExodus-2 (100.00 ng/mL), a concentration of 6.0 - 8.0 µg/mL of this antibody is required. ELISA: To detect hExodus-2 by direct ELISA (using 100 µL/well antibody solution) a concentration of at least 0.5 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of 0.2 - 0.4 ng/well of recombinant hExodus 2. Sandwich: To detect hExodus-2 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5 - 2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our Biotinylated Anti-Human Exodus-2 (XP-5128Bt) as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hExodus-2. Western Blot: To detect hExodus-2 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hExodus-2 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-hExodus-2 specific antibody was purified by affinity chromatography employing immobilized hExodus-2 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CCL21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human