To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 24
- Clonality:Polyclonal
- Gene ID:Q9JKC0
- Host:Goat
- Reactivity:Mouse
- Antigen symbol:Ccl24
- Conjugation:Biotin
- ELISA:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Ccl24
- Shipping temperature:RT
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant mEotaxin-2 (murine Eotaxin-2).
- Purification:Anti-mEotaxin-2 specific antibody was purified by affinity chromatography and then biotinylated.
- Cat. no.:10098-444
- Supplier No.:XP-5125BT
Specifications
About this item
Eotaxin is a 74 amino acid, eosinophil chemotactic CC chemokine originally found in bronchoalveolar lavage fluid from allergic inflammatory subjects. It is involved in regulating the recruitment and activation of inflammatory leukocytes, particularly eosinophils. Eotaxin binds and activates the CCR3 chemokine receptor, and may play a fundamental role in the development of allergic responses.
ELISA: To detect mEotaxin-2 by direct ELISA (using 100 µL/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 µg/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant mEotaxin-2. Sandwich: To detect mEotaxin-2 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25 - 1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Murine Eotaxin-2 (XP-5125) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant mEotaxin-2. Western Blot: To detect mEotaxin-2 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant mEotaxin-2 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Anti-mEotaxin-2 specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Ccl24
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse