To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) ligand 16
- Clonality:Polyclonal
- Gene ID:Q9H2A7
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CXCL16
- Conjugation:Biotin
- ELISA:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CXCL16
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hCXCL16 (Human CXCL16).
- Purification:Anti-hCXCL16 specific antibody was purified by affinity chromatography and then biotinylated.
- Cat. no.:10098-418
- Supplier No.:XP-5117BT
Specifications
About this item
ELISA:
To detect hCXCL16 by direct ELISA (using 100 ?L/well antibody solution) this antibody can be used at a concentration of 0.15 - 0.30 ?g/mL. Used in conjunction with compatible secondary reagents, allows the detection of at least 0.2 ng/well of recombinant hCXCL16.
Sandwich: To detect hCXCL16 by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our Polyclonal Anti-Human CXCL16 (XP-5117) as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hCXCL16.
Western Blot:
To detect hCXCL16 by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hCXCL16 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CXCL16
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human