To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 13 / adiponectin, C1Q and collagen domain containing
- Clonality:Monoclonal
- Clone:Sacha-2
- Gene ID:O75888 / Q15848
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Antigen symbol:TNFSF13 / ADIPOQ
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:TNFSF13 / ADIPOQ
- Storage buffer:PBS containing 0.02% sodium azide
- Storage temperature:APRIL antibody can be stored at 4 °C, stable for one year.
- Shipping temperature:4 °C
- Immunogen:APRIL (monoclonal Sacha-2) antibody was raised against recombinant human APRIL (aa. 105-250).
- Tested applications:Immunocytochemistry
- Purification:APRIL (monoclonal Sacha-2) antibody was purified from concentrated hybridoma tissue culture supernatant. APRIL (monoclonal Sacha-2) antibody was purified to 95% in SDS-PAGE.
- Cat. no.:10070-746
- Supplier No.:XA-1025
Specifications
About this item
APRIL antibody can be used to recognize human and mouse APRIL by ELISA (capture), Flow Cytometry, and Immunocytochemistry. Method: HEK-293T cells expressing surface membrane associated human APRIL (aa 92-233) or a mock control were injected in chicken liver and frozen in OCT. 8 ?m sections of injected frozen liver were fixed in acetone, washed in PBS and incubated with Sacha-2 at 10 ?g/mL for 1hr after blocking in PBS/1%BSA. Human APRIL was revealed with 3-amino-9-Ethyl-carbazole (AEC) following 45 min. incubation with goat anti-rat HRP. Method: HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mouse APRIL (thick line). Cells were incubated on ice for 30 min in 50 ?l FACS buffer (PBS, 5% Fetal calf serum, 0.02% sodium azide) containing 1 ?g/mL of Sacha-2 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry. This product is for research use only.
PBS containing 0.02% sodium azide
APRIL antibody can be stored at 4°C, stable for one year.
Type: Primary
Antigen: TNFSF13 / ADIPOQ
Clonality: Monoclonal
Clone: Sacha-2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse