To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 13
- Clonality:Monoclonal
- Clone:Aprily-8
- Gene ID:O75888
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:TNFSF13
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TNFSF13
- Storage buffer:PBS containing 0.02% sodium azide
- Storage temperature:APRIL antibody can be stored at 4 °C, stable for one year.
- Shipping temperature:4 °C
- Immunogen:APRIL (monoclonal Aprily-8) antibody was raised against recombinant human APRIL (aa. 93-233) expressed in bacteria.
- Tested applications:Immunocytochemistry
- Purification:APRIL (monoclonal Aprily-8) antibody was purified to >=95% in SDS-PAGE.
- Cat. no.:10070-736
- Supplier No.:XA-1020
Specifications
About this item
APRIL antibody can be used to recognize human APRIL by Flow Cytometry, immunocytochemistry and Western Blot. Method: 3 days after transfection of cells with the indicated constructs, cells were fixed with 4% formaldehyde 5 min at RT. After a wash in PBS, samples were dehydrated by washes in 60%, 80%, 90%, 100% EtOH and xylol. Cells were then dried and embedded in paraffin. Sections were cut, mounted on slides and dried overnight at 50°C. Slides were then successively washed 2x 10min in xylol, 2x 10min in 100% EtOH, and then treated 10 min in 100% MeOH/ 0.6% H202 to inhibit endogenous peroxidase. Samples were rehydrated by washes in 90%, 80%, 60% EtOH and PBS. After micro-wave treatment, slides were washed 3x in PBS, blocked with IgG, and incubated for 1 hour with 5 ?g/mL Aprily-8 or control mouse IgG (isotype control) in 1%BSA / 1x PBS for 1 hour. After PBS washes, samples were incubated with the secondary Ab for 1 hour, washed in PBS and revealed with StreptABComplex/HRP (Vector) and AEC. Method: HEK 293 cells were mock transfected or transfected with an expression plasmid coding for human APRIL. Cells (5x105) were incubated on ice for 30 min in 50 ?l FACS buffer (PBS, 5% fetal calf serum, 0.02% azide) containing 10 ?g/mL of Aprily-8 antibody. After washing in FACS buffer, FITC-conjugated antibody to mouse IgG was added. Cells were incubated on ice for 30 min, washed and analyzed by flow cytometry. Method: 10 ?g of protein was applied to the gel, and revealed with Aprily-8 (1 ?g/mL) and HRP-coupled anti-mouse secondary antibody (1:4,000).Note: The hAPRIL construct used in this experiment is an uncleavable fusion protein between hBAFF (aa 1-132) and hAPRIL (aa 93-233). This product is for research use only.
PBS containing 0.02% sodium azide
APRIL antibody can be stored at 4°C, stable for one year.
Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: Aprily-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human