To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 13
- Clonality:Monoclonal
- Clone:Aprily-1
- Gene ID:O75888
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:TNFSF13
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TNFSF13
- Storage buffer:PBS containing 0.02% sodium azide
- Storage temperature:APRIL antibody can be stored at 4 °C, stable for one year.
- Shipping temperature:4 °C
- Immunogen:APRIL (monoclonal Aprily-1) antibody was raised against recombinant human APRIL (aa. 93-233).
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:APRIL (monoclonal Aprily-1) antibody was purified to >=95% in SDS-PAGE.
- Cat. no.:10070-734
- Supplier No.:XA-1019
Specifications
About this item
APRIL antibody can be used to recognize human APRIL by Flow Cytometry (also works for intracellular FC staining) and Western Blot. Anti-APRIL (MAb Aprily-1) does not cross react with mouse APRIL. Method: HEK 293 cells were mock transfected or transfected with an expression plasmid enabling surface expression of hAPRIL. Cells (5x105) were incubated on ice for 30 min in 50 ?l FACS buffer (PBS, 5% Fetal calf serum, 0.02% azide) containing 10 ?g/mL of Aprily-1 antibody. After washing in FACS buffer, FITC-conjugated antibody to mouse IgG was added. Cells were incubated on ice for 30 min, washed and analyzed by flow cytometry. Method: Cell supernatants were first precleared with protein-A sepharose and then subjected to immunoprecipitation with rhBCMA:Fc and protein-A sepharose. Beads were washed twice with PBS. Immunoprecipitated materials were eluted with 2.5mM glycine pH 2.5 for 10 min, neutralized with Tris buffer and run on an SDS-PAGE gel and analyzed with 2 ?g/mL Aprily-1 by Western blot. Note: In some migrations endogenous soluble APRIL resolved as a doublet around 20kDa (dendritic cells: DC). rhsMegaAPRIL served as a positive control. Method: 3 HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mDR6 (thick line). Cells were incubated on ice for 30 min in 50 ?l FACS buffer (PBS, 5% Fetal calf serum, 0.02% azide) containing 1 ?g/mL of Luke-1 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry.
PBS containing 0.02% sodium azide
APRIL antibody can be stored at 4°C, stable for one year.
Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: Aprily-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human