To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor (ligand) superfamily, member 13
- Clonality:Monoclonal
- Clone:Sacha-1
- Gene ID:O75888
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Antigen symbol:TNFSF13
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:TNFSF13
- Storage buffer:PBS containing 0.02% sodium azide
- Storage temperature:APRIL antibody can be stored at 4 °C, stable for one year.
- Shipping temperature:4 °C
- Immunogen:APRIL (monoclonal Sache-1) antibody was raised against recombinant human APRIL (aa. 105-250).
- Tested applications:Immunocytochemistry
- Purification:APRIL (monoclonal Sache-1) antibody was purified to ≥95% in SDS-PAGE.
- Cat. no.:10070-724
- Supplier No.:XA-1014
Specifications
About this item
APRIL antibody can be used to recognize human and mouse APRIL by Flow Cytometry (excellent for mouse APRIL, also stains human APRIL), and Immunocytochemistry. Method: 3 days after transfection of cells with the indicated constructs, cells were fixed with acetone. Slides were blocked with IgG, and incubated for 1 hour with 5 ?g/mL Sacha-1 or control murine IgG (isotype control) in 1%BSA / 1x PBS for 1 hour. After washes wish PBS, samples were incubated with the secondary Ab for 1 hour, washed in PBS and revealed with StreptABComplex/HRP (Vector) and AEC. Method: 3 HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mAPRIL (thick line). Cells were incubated on ice for 30 min in 50 ?l FACS buffer (PBS, 5% Fetal calf serum, 0.02% azide) containing 1 ?g/mL of Sacha-1 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry.
PBS containing 0.02% sodium azide
APRIL antibody can be stored at 4°C, stable for one year.
Type: Primary
Antigen: TNFSF13
Clonality: Monoclonal
Clone: Sacha-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse