To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:v-myc avian myelocytomatosis viral oncogene homolog
- Clonality:Polyclonal
- Gene ID:P01106
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MYC
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MYC
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:60 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Myc (Ab-58) antibody was raised against a peptide sequence around aa.56~60 (L-P-T-P-P) derived from Human Myc.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:10068-788
- Supplier No.:79-281
Specifications
About this item
Myc proto-oncogene encodes nuclear DNA-binding phosphoproteins that are involved in the regulation of gene expression and DNA replication during cell growth and differentiation. Myc encodes a protein of 65 kDa which is expressed in almost all normal and transformed cells. The expression correlates with the proliferation state of the cells. Transcription is repressed in quiescent or terminally differentiated cells. Expression of Myc is generally induced after mitogenic stimulation of cells or serum induction. Myc therefore is an important positive regulator of cell growth and proliferation. Myc has been demonstrated also to be a potent inducer of apoptosis when expressed in the absence of serum or growth factors. Apoptosis may serve also as a protective mechanism to prevent tumorigenicity elicited by deregulated Myc expression. Sequences of the Myc oncogene have been highly conserved throughout evolution, from Drosophila to vertebrates
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat