To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:PHD finger protein 14
- Clonality:Polyclonal
- Gene ID:O94880
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PHF14
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PHF14
- Storage buffer:PHF14 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 98 kDa Observed: 100kDa
- Storage temperature:PHF14 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:PHF14 antibody was raised against a 19 amino acid peptide near the amino terminus of human PHF14. The immunogen is located within amino acids 210 - 260 of PHF14 .
- Purification:PHF14 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-978
- Supplier No.:7485
Specifications
About this item
PHF14, a novel plant homology domain (PHD) transcription factor, controls the proliferation of mesenchymal cells by regulating PDGFR alpha expression in embryonic development. PHD finger transcription factors are concerned with transcriptional regulation by interacting with modified histones (2), and dysregulation of several genes encoding PHD finger transcription factors, such as ING1 and RBP2, has been found in human cancer diseases suggesting these factors have an important role for cancer cell proliferation. PHF14 is a potential target for new treatments of lung fibrosis (1, 3).
PHF14 antibody can be used for detection of PHF14 by Western blot at 1 - 2 µg/ml.
PHF14 Antibody is affinity chromatography purified via peptide column.
PHF14 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PHF14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat