To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:T-box 21
- Clonality:Polyclonal
- Gene ID:Q9UL17
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:TBX21
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TBX21
- Storage buffer:TBX21 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 59 kDa Observed: 55kDa
- Storage temperature:TBX21 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:TBX21 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TBX2. The immunogen is located within the last 50 amino acids of TBX21 .
- Purification:TBX21 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-972
- Supplier No.:7477
Specifications
About this item
TBX21 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box (1,2). Members of this family include transcription factors involved in the regulation of developmental processes. Studies in mouse and humans show that TBX21 is a Th1 cell-specific transcription factor that controls the expression of the hallmark Th1 cytokine, interferon-gamma (1,3). Expression of the human ortholog also correlates with interferon-gamma expression in Th1 and natural killer cells, suggesting a TBX21 may play a role in initiating Th1 lineage development from naive Th precursor cells..
TBX21 antibody can be used for detection of TBX21 by Western blot at 1 - 2 µg/ml.
TBX21 Antibody is affinity chromatography purified via peptide column.
TBX21 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TBX21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse