To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:mucin 1, cell surface associated
- Clonality:Polyclonal
- Gene ID:P15941
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:MUC1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MUC1
- Storage buffer:MUC1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 53 (+ multiple others) kDa Observed:42, 120 kDa
- Storage temperature:MUC1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:MUC1 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MUC1. The immunogen is located within the last 50 amino acids of MUC1 .
- Purification:MUC1 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-952
- Supplier No.:7451
Specifications
About this item
Mucin 1 (MUC1) is membrane-bound protein member of the mucin family. Mucins are O-glycosylated proteins that play an essential role in forming protective mucous barriers on epithelial surfaces and also play a role in intracellular signaling. MUC1 is expressed on the apical surface of epithelial cells that line the mucosal surfaces of many different tissues including lung, breast stomach and pancreas. MUC1 is proteolytically cleaved into alpha and beta subunits that form a heterodimeric complex. The N-terminal alpha subunit functions in cell-adhesion and the C-terminal beta subunit is involved in cell signaling. Overexpression, aberrant intracellular localization, and changes in glycosylation of this protein have been associated with carcinomas.
MUC1 antibody can be used for detection of MUC1 by Western blot at 1 - 2 µg/ml.
MUC1 Antibody is affinity chromatography purified via peptide column.
MUC1 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse