To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 33
- Clonality:Polyclonal
- Gene ID:Q9UPN9
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TRIM33
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TRIM33
- Storage buffer:TRIM33 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 70, 124 kDa Observed: 70 kDa
- Storage temperature:TRIM33 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:TRIM33 antibody was raised against an 18 amino acid peptide near the center of human TRIM33. The immunogen is located within amino acids 580 - 630 of TRIM33.
- Purification:TRIM33 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-916
- Supplier No.:7385
Specifications
About this item
TRIM33 Antibody: TRIM24 (TIF1alpha), TRIM28 (TIF1beta), and TRIM33 (TIF1gamma) are three related cofactors belonging to the tripartite motif superfamily that interact with distinct transcription factors. The structure of TRIM33 is similar to TRIM24 and TRIM28, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TRIM33 functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells. It interacts with the Smad2/3 transcription factor in hematopoietic, mesenchymal, and epithelial cell types to mediate different transcriptional effects in response to TGF-beta. It acts as an E3 ubiquitin-protein ligase and promotes SMAD4 ubiquitination.
TRIM33 antibody can be used for detection of TRIM33 by Western blot at 1 - 2 ?g/mL.
TRIM33 Antibody is affinity chromatography purified via peptide column.
TRIM33 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TRIM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat