To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding)
- Clonality:Polyclonal
- Gene ID:Q7Z429
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:GRINA
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GRINA
- Storage buffer:GRINA Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 41 kDa
- Storage temperature:GRINA antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Rabbit polyclonal GRINA antibody was raised against an 18 amino acid peptide near the center of human GRINA. The immunogen is located within amino acids 110 - 160 of GRINA.
- Purification:GRINA Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-774
- Supplier No.:7147
Specifications
About this item
GRINA Antibody: The transmembrane BAX inhibitor motif (TMBIM) family of proteins includes the founder member TMBIM6/BI-1, TMBIM1/RECS1 (responsive to centrifugal force and shear stress gene 1 protein), TMBIM2/LFG (life guard), TMBIM3/GRINA (glutamate receptor ionotropic NMDA protein 1), TMBIM4/GAAP (Golgi anti-apoptotic-associated protein), and TMBIM5/GHTIM (growth hormone-inducible transmembrane protein). They are highly conserved in mammals and zebrafish and contain a conserved BAX inhibitor-1 motif. GRINA is expressed in the brain and is a potential apoptotic regulator.
GRINA antibody can be used for detection of GRINA by Western blot at 1 - 2 ?g/mL.
GRINA Antibody is affinity chromatography purified via peptide column.
GRINA Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GRINA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human