To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:signal peptide, CUB and EGF-like domain-containing protein 3 precursor
- Clonality:Polyclonal
- Gene ID:Q8IX30
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:SCUBE3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:SCUBE3
- Storage buffer:SCUBE3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 109 kDa
- Storage temperature:SCUBE3 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:SCUBE3 antibody was raised against a 15 amino acid peptide near the center of human SCUBE3. The immunogen is located within amino acids 440 - 490 of SCUBE3.
- Purification:SCUBE3 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10061-904
- Supplier No.:5847
Specifications
About this item
SCUBE3 Antibody: SCUBE3 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and is highly expressed in primary osteoblasts and bones, and to a lesser extent in heart (1,2). Other studies have shown that overexpression of SCUBE3 in mice induced cardiac hypertrophy, suggesting that it may also play a role in the regulation of cardiac growth. SCUBE3 has been shown to be an endogenous TGF-beta receptor ligand (3,4) and is thought to promote lung cancer cell mobility and invasiveness. In lung cancer cells, the secreted SCUBE3 protein was cleaved by MMP2 and MMP9, allowing the activation of the TGF-beta receptor, the increase of Smad2/3 transcriptional activity and the upregulation of expression of proteins such as TGF-beta1, VEGF, Snail, and Slug.
SCUBE3 antibody can be used for detection of SCUBE3 by immunohistochemistry at 5 ?g/mL.
SCUBE3 Antibody is affinity chromatography purified via peptide column.
SCUBE3 Antibody is supplied in PBS containing 0.02% sodium azide.
SCUBE3 antibody can be stored at 4°C for three months and -20°C, stable for up to one year.
Type: Primary
Antigen: SCUBE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human