To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta
- Clonality:Polyclonal
- Gene ID:P35213
- Host:Rabbit
- Reactivity:Zebrafish,Xenopus,Sheep,Chicken,Human,Bovine,Dog,Rat,Mouse
- Antigen symbol:Ywhab
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Ywhab
- Modification:Ser 58
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:29
- Storage temperature:14-3-3 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping temperature:4 °C
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser58 of rat 14-3-3 protein.
- Purification:Affinity Purified
- Cat. no.:10075-464
- Supplier No.:50-231
Specifications
About this item
14-3-3 proteins are a family of highly conserved proteins that appear to have multiple roles in cell signaling (Bridges and Moorhead, 2005). The proteins are abundantly expressed in the brain and have been detected in the cerebrospinal fluid of patients with different neurological disorders (Berg et al., 2003). 14-3-3 proteins bind protein ligands that are typically phosphorylated on
serine or threonine residues and regulate the functions of these binding partners by a number of different mechanisms (Silhan et al., 2004; Dougherty and Morrison, 2004). The14-3-3 proteins affect a diverse array of cellular processes including the cell cycle and transcription, signal transduction and intracellular trafficking. These functions of 14-3-3 proteins are facilitated by, if not dependent on, its dimeric structure. Recent work has demonstrated that the dimeric status of the 14-3-3 protein is regulated by site-specific serine phosphorylation (Woodcock et al., 2003).
The antibody has been directly tested for reactivity in Western blots with human and rat tissue. It is anticipated that the antibody will react with bovine, canine, chicken, mouse, non-human primates, sheep, Xenopus and zebra fish based on the fact that these species have 100% homology with the amino acid sequence used as antigen for this antibody.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Ywhab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Sheep, Bovine, Chicken, Xenopus