To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:retinoid X receptor, gamma
- Clonality:Monoclonal
- Clone:1373
- Gene ID:P48443
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Antigen symbol:RXRG
- Conjugation:Unconjugated
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RXRG
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:48
- Storage temperature:Retinoid X Receptor antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping temperature:4 °C
- Immunogen:Hinge peptide from human retinoid X receptor, y-isotype.
- Purification:Protein G purified
- Cat. no.:10075-642
- Supplier No.:50-174
Specifications
About this item
Retinoic acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXRalpha, beta and gamma) preferentially bind 9-cis-RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).
The antibody has been directly tested for reactivity in Western blots in human tissues. It is anticipated that the antibody will also work with bovine, mouse, non-human primate and rat tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Protein G purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RXRG
Clonality: Monoclonal
Clone: 1373
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine