To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:calcitonin-related polypeptide alpha
- Clonality:Monoclonal
- Clone:4901
- Gene ID:P01256
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat
- Antigen symbol:Calca
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Calca
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Shipping temperature:4 °C
- Immunogen:Rat alpha-CGRP
- Tested applications:Immunocytochemistry
- Purification:Protein G purified
- Cat. no.:10075-404
- Supplier No.:50-151
Specifications
About this item
Protein G purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: Calca
Clonality: Monoclonal
Clone: 4901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat