To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, ionotropic, N-methyl D-aspartate 2A
- Clonality:Polyclonal
- Gene ID:Q00959
- Host:Rabbit
- Reactivity:Rat,Mouse
- Antigen symbol:Grin2a
- Conjugation:Unconjugated
- Size:0.1 mL
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Grin2a
- Modification:Tyr 1325
- Storage buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular weight:180
- Shipping temperature:4 °C
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Tyr1325 of rat NMDA NR2A.
- Purification:Affinity Purified
- Cat. no.:10075-638
- Supplier No.:50-121
Specifications
About this item
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: Grin2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat