To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:cyclin-dependent kinase 2
- Clonality:Monoclonal
- Gene ID:P24941
- Host:Mouse
- Isotype:IgG
- Reactivity:Xenopus,Human,Mouse
- Antigen symbol:CDK2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CDK2
- Storage buffer:0.1M Tris-glycine, pH 7.4, 0.15M sodium chloride, 0.05% sodium azide, before the addition of glycerol to 30%
- Storage temperature:Store CDK2 antibody at 4 °C or –20 °C. As with all antibodies avoid freeze/thaw cycles.
- Concentration:0.85 mg/mL
- Shipping temperature:4 °C
- Immunogen:CDK2 monoclonal antibody was raised against recombinant CDK2 (Human).
- Purification:Protein G Column
- Cat. no.:10079-340
- Supplier No.:49-958
Specifications
About this item
Cyclin-dependent kinase 2 (CDK2), a CDC2/CDK-type protein kinase, regulates cell-cycle transition from G1 to S phase. CDK2 activation requires formation of a heterodimer with cyclin A, D, or E and phosphorylation by CAK. CDK2 inhibition results from association with p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). Two alternatively spliced variants and multiple transcription initiation sites have been reported. Inhibition of CDK2 has been suggested to help prevent chemotherapy-induced alopecia in cancer patients.
CDK2 antibody can be used in immunohistochemistry starting at 10 ?g/mL.
Protein G Column
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CDK2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Xenopus, Human, Mouse