To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:peroxisome proliferator-activated receptor gamma
- Clonality:Polyclonal
- Gene ID:P37231
- Host:Rabbit
- Reactivity:Mouse,Duck,Hamster,Rabbit,Squirrel,Boar,Macaque,Orangutan,Human,Guinea pig,Dog,Mink,Rat
- Antigen symbol:PPARG
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PPARG
- Storage buffer:0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, 0.01% sodium azide.
- Storage temperature:Store PPARG antibody at 4 °C or –20 °C. As with all antibodies avoid freeze/thaw cycles.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:PPARG antibody was raised against amino acids from the C-Terminus of PPARG (Human).
- Tested applications:Immunoaffinity Chromatography
- Purification:Immunoaffinity Chromatography
- Cat. no.:10076-412
- Supplier No.:49-549
Specifications
About this item
Since their discovery in the early 1990's, the peroxisome proliferator activated receptors (PPARs) have attracted significant attention. This is primarily because PPARs serve as receptors for two very important classes of drugs: the hypolipidemic fibrates and the insulin sensitizing thiazolidinediones. Peroxisome proliferators are non-genotoxic carcinogens that are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed PPARs. Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Upon binding fatty acids or hypolipidemic drugs, PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate the expression of target genes. There are 3 known subtypes of PPARs: PPAR-alpha, PPAR-delta and PPAR-gamma. Mostly target genes are involved in the catabolism of fatty acids. Conversely, PPAR-gamma is activated by peroxisome proliferators such as prostaglandins, leukotrienes and anti-diabetic thiazolidinediones and affects the expression of genes involved in the storage of the fatty acids. PPAR-gamma may also be involved in adipocyte differentiation. It has also been shown that PPARs can induce transcription of acyl coenzyme A oxidase and cytochrome P450 through interaction with specific response elements.
PPARG antibody can be used in Western Blot starting at 1:200 - 1:1000, and immunohistochemistry starting at 1:50.
0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PPARG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Boar, Dog, Duck, Guinea pig, Hamster, Human, Macaque, Mink, Mouse, Orangutan, Rabbit, Rat, Squirrel