To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:proliferating cell nuclear antigen
- Clonality:Monoclonal
- Clone:[PC10]
- Gene ID:P12004
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Antigen symbol:PCNA
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PCNA
- Storage buffer:Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.
- Storage temperature:Store PCNA antibody at 4 °C. Do not freeze.
- Concentration:0.5 mg/mL
- Shipping temperature:4 °C
- Immunogen:PCNA monoclonal antibody was raised against recombinant of PCNA (Human).
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Affinity Chromatography
- Cat. no.:10078-614
- Supplier No.:49-126
Specifications
About this item
PCNA antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.
Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.
Store PCNA antibody at 4 °C. Do not freeze.
Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: [PC10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse