To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:replication protein A3, 14kDa
- Clonality:Monoclonal
- Clone:[111]
- Gene ID:P35244
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Antigen symbol:RPA3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RPA3
- Storage buffer:PBS.
- Storage temperature:Short term: 4 °C. Long term: Store at –20 °C. Avoid freeze-thaw cycles.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Replication Protein A3 monoclonal antibody was raised against full length Replication Protein A3 expressed in E. coli (Human).
- Purification:Protein G Column
- Cat. no.:10074-446
- Supplier No.:48-861
Specifications
About this item
Replication protein A (RPA) is a single-stranded DNA binding protein. Human RPA is a heterotrimeric protein containing subunits of 70, 32 and 14 kDa. This protein complex is highly conserved in eukaryotes and is essential in DNA replication, homologous recombination and nucleotide excision repair. RPA has been shown to be a target for DNA-dependent protein kinase (DNA-PK) ataxia telangiectasia-mutated gene (ATM) and a cyclin dependent protein kinase.
Replication Protein A3 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.
Protein G Column
PBS.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RPA3
Clonality: Monoclonal
Clone: [111]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse