To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tyrosyl-DNA phosphodiesterase 2
- Clonality:Monoclonal
- Clone:G8
- Gene ID:O95551
- Host:Mouse
- Isotype:IgG1
- Reactivity:Xenopus,Chicken,Human,Rat,Mouse
- Antigen symbol:TDP2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TDP2
- Storage buffer:PBS
- Storage temperature:Ski monoclonal antibody can be stored at 4 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:Recombinant v-ski.
- Tested applications:Immunocytochemistry
- Purification:Protein G-purified
- Cat. no.:10070-980
- Supplier No.:47-141
Specifications
About this item
The ski oncogene family (ski and sno) encodes transcriptional regulators that can affect oncogenic transformation and cellular differenti-ation. ski and sno function as co-activators or co-repressors, modulating transcription by inter-acting with transcription factor complexes. ski interacts with members of the Nuclear Factor I (NFI) family, the retinoic acid receptor, and the nuclear hormone co-regulators NCoR and NCoA62/skip.
Protein G-purified
PBS
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TDP2
Clonality: Monoclonal
Clone: G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken, Xenopus