To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ligase III, DNA, ATP-dependent
- Clonality:Polyclonal
- Gene ID:P49916
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:LIG3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:LIG3
- Storage buffer:PBS
- Storage temperature:ESE-1 antibody can be stored at 4 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:N-terminal sequence of human ESE-1 protein.
- Cat. no.:10069-718
- Supplier No.:47-131
Specifications
About this item
Epithelium-specific gene regulation plays acritical role during embryogenesis, and the epithelial cell lineage is the first differentiated cell type to appear after fertilization. Until recently, very few epithelial cell-restricted transcription factors had been described. Members of the ets transcription factor/oncogene family have been implicated in human carcinogenesis following the discovery of several distinctchromosomal translocations involving members of the ets family in different types of cancer. ets factors play key roles intranscriptional control of stringently regulated genes, such as genes involved in tissue development, differentiation,angiogenesis, cell cycle control and cellproliferation. ESE-1 is the first member ofthe ets family that was found to be exclusively expressed and inducible inepithelial cells.
PBS
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: LIG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human