To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:LIM domain binding 3
- Clonality:Polyclonal
- Gene ID:O75112
- Host:Goat
- Reactivity:Human
- Antigen symbol:LDB3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:LDB3
- Storage buffer:ZASP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:ZASP antibody was raised against a 12 amino acid synthetic peptide near the internal region of ZASP.
- Purification:ZASP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-902
- Supplier No.:46-608
Specifications
About this item
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Experiments gave bands at approximately 80-90 kDa and 30-35 kDa in human muscle lysates after 0.01 ?g/mL antibody staining. These bands correspond to earlier findings in literature with different antibodies (Huang et al J Biol Chem. 2003 Feb 28;278 (9) :7360-5; PMID: 12499364). This protein has a calculated MW of 77.1 kDa according to NP_009009.1. Recommended concentration: 0.01-0.03 ?g/mL. Immunohistochemistry: In paraffin embedded human skeletal muscle shows staining of the Z lines in the fibres. Recommended concentration, 3-6 ?g/mL.
ZASP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: LDB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human