To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:signal-induced proliferation-associated 1
- Clonality:Polyclonal
- Gene ID:Q96FS4
- Host:Goat
- Reactivity:Human
- Antigen symbol:SIPA1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SIPA1
- Storage buffer:SIPA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:SIPA1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of SIPA1.
- Purification:SIPA1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-720
- Supplier No.:46-371
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Approximately 150 kDa band observed in human lysates of EBV-immortilized lymphoblastoid (calculated MW of 112 kDa according to NP_694985.28 and NP_006738.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Kurachi et al, J Biol Chem. 1997 Oct 31;272 (44) :28081-8; PMID: 9346962). Recommended concentration: 1-3 ?g/mL. an additional band of 55-60 kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal. We call for caution when this antibody is used for other assays than Western Blot.
SIPA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SIPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human