To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SECIS binding protein 2
- Clonality:Polyclonal
- Gene ID:Q96T21
- Host:Goat
- Reactivity:Human,Mouse
- Antigen symbol:SECISBP2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SECISBP2
- Storage buffer:SBP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:SBP2 antibody was raised against a 13 amino acid synthetic peptide near the internal region of SBP2.
- Tested applications:Immunocytochemistry
- Purification:SBP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-668
- Supplier No.:46-339
Specifications
About this item
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 120 kDa band detected in HFF cell lysates. Data kindly provided by Laura Papp and Kum Kum Khanna. The observed molecular weight corresponds to earlier findings in literature with different antibodies (Papp et al, Mol Cell Biol. 2006 Jul;26 (13) :4895-910; PMID: 16782878). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human testis shows granular cytoplasm staining in Leydig cells. Recommended concentration, 3 ?g/mL.
SBP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SECISBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse