To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 40
- Clonality:Polyclonal
- Gene ID:Q6P9F5
- Host:Goat
- Reactivity:Human
- Antigen symbol:TRIM40
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TRIM40
- Storage buffer:RNF35 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:RNF35 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of RNF35.
- Purification:RNF35 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-624
- Supplier No.:46-310
Specifications
About this item
ELISA: Antibody detection limit dilution >1:32000. Western Blot: Approximately 45 kDa band observed in human brain lysates at 0.5-1 ?g, this band was successfully blocked by incubation with the immunizing peptide. Please note that the band we observe is different from the predicted size of approximately 26 kDa according to NP_619645. Approximately 26 kDa band observed in human placenta lysates (calculated MW of 26.1 kDa according to NP_619645.1). Recommended concentration: 0.5-2 ?g/mL.
RNF35 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TRIM40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human