To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:RAB11 family interacting protein 1 (class I)
- Clonality:Polyclonal
- Gene ID:Q6WKZ4
- Host:Goat
- Reactivity:Human
- Antigen symbol:RAB11FIP1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RAB11FIP1
- Storage buffer:RAB11FIP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:RAB11FIP1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of RAB11FIP1.
- Purification:RAB11FIP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-554
- Supplier No.:46-257
Specifications
About this item
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 110 kDa, 50 kDa and 38 kDa in human breast lysate after 1 ?g/mL antibody staining.0 kDa according to NP_079427.3, and 137 kDa according to NP_001002814.1. The detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Please note that an additional 55 kDa was only partly blocked by the immunizing peptide, this band may not be specific. Approximately 80 kDa band observed in HeLa lysate (calculated MW of 71.0 kDa according to NP_079427.3). The observed molecular weight of 80 kDa corresponds to earlier findings in literature with different antibodies (Lindsay et al, J Biol Chem. 2002 Apr 5;277(14):12190-9. PMID 11786538.). Recommended concentration: 0.1-0.3 ?g/mL.
RAB11FIP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RAB11FIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human