To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:phosphodiesterase 4D, cAMP-specific
- Clonality:Polyclonal
- Gene ID:Q08499
- Host:Goat
- Reactivity:Human,Mouse
- Antigen symbol:PDE4D
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PDE4D
- Storage buffer:PDE4D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:PDE4D antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of PDE4D.
- Tested applications:EIA
- Purification:PDE4D antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10116-402
- Supplier No.:46-151
Specifications
About this item
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 70+80+90 kDa bands observed in A549 lysate . In transfected COS1 transiently expressing PDE4D1 and PDE4D3 bands of approximately 68 kDa and 95 kDa are observed. These bands are not observed in the untransfected COS1. In transfected HEK293 transiently expressing PDE4D2 multi bands from Approximately 55 kDa to 80 kDa are observed. These bands are not observed in the non-transfected HEK293. Calculated MW of 91.1 kDa according to PDE4D1 (NP_001098101.1), 76.5 kDa according to PDE4D2 (NP_006194.2) and 84.7 kDa according to PDE4D3 (NP_001159371.1). Recommended concentration: 0.1-0.3 ?g/mL.
PDE4D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: PDE4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse